Learn More
Med Vet International 1/2cc 28g x 1 1/2in. Insulin Syringe, 500/Case
Catalog No. 502092934
Encompass_Preferred
In convenient packages of 10. Quality compared to national leading brands Color-Coded to Gauge Easy-to-read information on each side of the box Needles are made with Surgical Steel Triple-Bevel Cut for a Sharper Point Thin Wall Cannulas for an Easier Flow Electro-polished to Remove Burrs for a Smoother Injection Film Coated for Maximum Comfort
Catalog No. 50-209-2934 Supplier Med Vet International Supplier No. 828555X5
In convenient packages of 10. Quality compared to national leading brands Color-Coded to Gauge Easy-to-read information on each side of the box Needles are made with Surgical Steel Triple-Bevel Cut for a Sharper Point Thin Wall Cannulas for an Easier Flow Electro-polished to Remove Burrs for a Smoother Injection Film Coated for Maximum Comfort