Learn More
Med Vet International Exel Syringe & Needle, .5mL, Permanent Needle, 28G X 1/2in, Insulin, 100/BX, 26026
Catalog No. 502092933
Encompass_Preferred
In convenient packages of 10. Exel International .5mL Syringe comes with a clear barrel and easy to read graduation marks. This syringe contains a permanently attached needle. This Exel International 26026 Syringe & Needle Combo comes with a 28G X 1/2' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This is an insulin syringe. It is used to inject insulin in the subcutaneous fat. This quantity is 100/BX
Catalog No. 50-209-2933 Supplier Med Vet International Supplier No. 26026
In convenient packages of 10. Exel International .5mL Syringe comes with a clear barrel and easy to read graduation marks. This syringe contains a permanently attached needle. This Exel International 26026 Syringe & Needle Combo comes with a 28G X 1/2' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This is an insulin syringe. It is used to inject insulin in the subcutaneous fat. This quantity is 100/BX