Learn More
Med Vet International Exel Syringe & Needle, .3mL, Permanent Needle, 29G X 1/2in, Insulin, 100/Box, 26018
Catalog No. 502092937
Encompass_Preferred
In convenient packages of 10. Exel International .3mL Syringe comes with a clear barrel and easy to read graduation marks. This syringe contains a permanently attached needle. This Exel International 26018 Syringe & Needle Combo comes with a 29G X 1/2in. Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This is an insulin syringe. It is used to inject insulin in the subcutaneous fat. This quantity is 100/BX
Catalog No. 50-209-2937 Supplier Med Vet International Supplier No. 26018
In convenient packages of 10. Exel International .3mL Syringe comes with a clear barrel and easy to read graduation marks. This syringe contains a permanently attached needle. This Exel International 26018 Syringe & Needle Combo comes with a 29G X 1/2in. Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This is an insulin syringe. It is used to inject insulin in the subcutaneous fat. This quantity is 100/BX