Learn More
Trajan Scientific SYRNG 10UL FXDNDL VPKIOF 10/PK
SDP
Catalog No. 502530406
Encompass_Preferred
SGE Series II Standard Fixed Needle Syringes, Volume: 10 uL, Needle Gauge: 26 G, Includes: Plunger protection, Length Needle: 50 mm, 1.96 in., Accuracy: +/-1%, Connection Type: Fixed, Diameter Outer: 0.25 in., 6.5 mm, Diameter Outer Needle: 0.01 in., 0.47 mm
Catalog No. 50-253-0406 Supplier Trajan Scientific Supplier No. 002033
SGE Series II Standard Fixed Needle Syringes, Volume: 10 uL, Needle Gauge: 26 G, Includes: Plunger protection, Length Needle: 50 mm, 1.96 in., Accuracy: +/-1%, Connection Type: Fixed, Diameter Outer: 0.25 in., 6.5 mm, Diameter Outer Needle: 0.01 in., 0.47 mm