Learn More
Tisch Scientific Tisch Scientific Syringe Filter, Glass Fiber, 1.0 µm, 30 mm, Luer-Lok/Luer Slip, Nonsterile, 100 Pack, SF18163
SDP
Supplier:  Tisch Scientific SF18163
Encompass
Tisch glass fiber syringe filters are ideal for the filtration of extremely viscous samples which could clog other standard syringe filters. These hydrophilic syringe filters are known for their excellent compatibility with organic solvents, strong acids and bases
Catalog No. NC1616133