Learn More
Cayman Chemical (−)-Syringaresinol di-O-glucoside, Purity: ≥98%, CAS: 66791-77-3, 10 mg
SDP
Catalog No. 502910069
Encompass_Preferred
A phenolic glucoside with in vitro neuroprotective activity; reduces glutamate-induced neurotoxicity by 19.3% in PC12 cells at 10 µM
Catalog No. 50-291-0069 Supplier Cayman Chemical Supplier No. 3667210 MG
A phenolic glucoside with in vitro neuroprotective activity; reduces glutamate-induced neurotoxicity by 19.3% in PC12 cells at 10 µM