Learn More
Chemglass Life Sciences Syringe, 5mL Polypropylene, Luer Lock
SDP
Catalog No. 50872581
Encompass_Preferred
Syringe, 5mL Polypropylene, Luer Lock. All polypropylene syringes with a Luer Lock. The plastic plunger has a smooth draw and positive safety stop to prevent accidental spills. Syringes are individually sterile strip packed.
Catalog No. 50-872-581 Supplier Chemglass Life Sciences Supplier No. CG308102
Syringe, 5mL Polypropylene, Luer Lock. All polypropylene syringes with a Luer Lock. The plastic plunger has a smooth draw and positive safety stop to prevent accidental spills. Syringes are individually sterile strip packed.