Learn More
Med Vet International Exel Tuberculin Syringe & Needle, 1mL, Luer Slip, 26G X 1/2in, 100/BX, 26043
Catalog No. 502092948
Encompass_Preferred
Exel International 1mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer Lock tip of this 1mL syringe fits conventional and safety needles. This Exel International 26043 Syringe & Needle Combo comes with a 26G X 1/2' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This is a tuberculin syringe. They are small syringes with fine needles used to administer medication under the skin and perform tuberculosis tests. This quantity is 100/BX
Catalog No. 50-209-2948 Supplier Med Vet International Supplier No. 26043EX
Exel International 1mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer Lock tip of this 1mL syringe fits conventional and safety needles. This Exel International 26043 Syringe & Needle Combo comes with a 26G X 1/2' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This is a tuberculin syringe. They are small syringes with fine needles used to administer medication under the skin and perform tuberculosis tests. This quantity is 100/BX