Learn More
Med Vet International Exel Syringe & Needle, .5mL, Permanent Needle, 29G X 1/2in, Insulin, Vet Use, 100/BX, 26036
Catalog No. 502092959
Encompass_Preferred
In convenient packages of 10. Exel International .5mL Syringe comes with a clear barrel and easy to read graduation marks. This syringe contains a permanently attached needle. This Exel International 26036 Syringe & Needle Combo comes with a 29G X 1/2' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This is an insulin syringe. It is used to inject insulin in the subcutaneous fat. This quantity is 100/BX
Catalog No. 50-209-2959 Supplier Med Vet International Supplier No. 26036
In convenient packages of 10. Exel International .5mL Syringe comes with a clear barrel and easy to read graduation marks. This syringe contains a permanently attached needle. This Exel International 26036 Syringe & Needle Combo comes with a 29G X 1/2' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This is an insulin syringe. It is used to inject insulin in the subcutaneous fat. This quantity is 100/BX