Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siemens Healthineers Sysmex™ CA-660 Hemostasis System

Catalog No. 23044750 Shop All Siemens Healthineers Products
Change view
Click to view available options
Capacity:
60 Reaction Tubes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
23-044-750 60 Reaction Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-044-750 Supplier Siemens Healthineers Supplier No. 11694252

This product is not orderable online. For ordering, please contact your Fisher Healthcare Sales Representative.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The CA-600 Series Systems build upon the proven technology and reliability of the Sysmex™ CA-500 Series Systems, thus offering peace of mind and easy integration into your laboratory.

  • Two system options to meet your lab’s needs
    • CA-620 for clotting detection
    • CA-660 for clotting, chromogenic, and immunologic detection
  • Hand-held 2D bar-code reader for safe and convenient data entry.
  • Tilted color LCD touch screen for easy-to-use system handling.
  • Sample bar-code reader provides simplified sample ID processing and enables
    security and integrity of patient records.

Measurement

  • Principle
    • Photo-optical, continuous, sequential
  • Method/channels
    • Clotting/4, chromogenic, and immunologic
  • Source lamp
    • Clotting – 4 LEDs with 660 nm/wavelength
    • Chromogenic – 1 LED with 405 nm/wavelength
    • Immunologic – 1 LED with 575 nm/wavelength
TRUSTED_SUSTAINABILITY

Specifications

Type Hemostasis System
For Use With (Application) Clotting, Chromogenic and Immunologic Detection
Sample Size 60 Reaction Tubes
Throughput PT 60 tests/hour; D-Dimer 16 tests/hour; PT, APTT, Fibrinogen 48 simultaneous tests/hour; PT/APTT/Fibrinogen/AT 40 simultaneous tests/hour; PT/APTT/Fib/D-Dimer 32 simultaneous tests/hour
Capacity 60 Reaction Tubes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.