Learn More
Sigma Aldrich Fine Chemicals Biosciences System 45TM vessel with th1EA
Catalog No. 502906168
Encompass
Cylindrical style vessel has a 2Amm PTfE valve at the top with a GL 18 threaded side port. The 32Amm threaded bottom valve assembly incorporates System 45(TM) technology. The coarse fritted disc is easily removed from the PTfE adapter for cleaning and/or replacement. PTfE top and bottom valves require 1/4-turn to open or close. Valve stems have a 1/4-28 internal thread in the stem which allows a variety of adapters to be used. for male 1/4-28 and male Luer adapters see Cat. No. Z417416 and Z417424.
Catalog No. 50-290-6168 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z4188621EA
Cylindrical style vessel has a 2Amm PTfE valve at the top with a GL 18 threaded side port. The 32Amm threaded bottom valve assembly incorporates System 45(TM) technology. The coarse fritted disc is easily removed from the PTfE adapter for cleaning and/or replacement. PTfE top and bottom valves require 1/4-turn to open or close. Valve stems have a 1/4-28 internal thread in the stem which allows a variety of adapters to be used. for male 1/4-28 and male Luer adapters see Cat. No. Z417416 and Z417424.