Learn More
MERCEDES SCIENTIFIC System Blade Removal QuickSmart - Safe,quick & easy to use. 100 blade capcity. Single-handed use keeping hands out of harms way. Self-contained waste disposal system. Use with blades from handles #3,4,5,7 & 9.
Catalog No. NC3450210
Encompass
System Blade Removal QuickSmart - Safe,quick & easy to use. 100 blade capcity. Single-handed use keeping hands out of harms way. Self-contained waste disposal system. Use with blades from handles #3,4,5,7 & 9. Blade fully contained upon removal, eliminating injuries to downstream staff.
This item is not returnable. View return policy
Catalog No. NC3450210 Supplier MERCEDES SCIENTIFIC Supplier No. QST QFRGEN
System Blade Removal QuickSmart - Safe,quick & easy to use. 100 blade capcity. Single-handed use keeping hands out of harms way. Self-contained waste disposal system. Use with blades from handles #3,4,5,7 & 9. Blade fully contained upon removal, eliminating injuries to downstream staff.