Learn More
Quanterix Inc SYSTEM BUFFER 1
SDP
Catalog No. NC0621647
Encompass
System Buffer 1, (2 Pack)- 2x (5L) System Buffer 1 specifically formulated to optimize performance of assays on the Simoa HD-1 Analyzer
Catalog No. NC0621647 Supplier Quanterix Inc Supplier No. MK20032
System Buffer 1, (2 Pack)- 2x (5L) System Buffer 1 specifically formulated to optimize performance of assays on the Simoa HD-1 Analyzer