Learn More
BIOMEDIX SYSTEM SURE REAGENT, liquid-stable luciferase/luciferin, ATP detection, compatible with UltraSnap and AquaSnap devices, response time 15 sec
Catalog No. NC0512092
Encompass
SYSTEM SURE REAGENT, liquid-stable luciferase/luciferin, ATP detection, compatible with UltraSnap and AquaSnap devices, response time 15 sec
Catalog No. NC0512092 Supplier BIOMEDIX Supplier No. 1216456
SYSTEM SURE REAGENT, liquid-stable luciferase/luciferin, ATP detection, compatible with UltraSnap and AquaSnap devices, response time 15 sec