Learn More
Aviva Systems Biology Corp SYT11 Antibody - N-terminal region (ARP59473_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59473_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SYT11. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human SYT11. Alternate names: DKFZp781D015, KIAA0080, MGC10881, MGC17226, SYT12, sytXIApplicationWB. Species reactivity: Guinea Pig, Human, Mouse
Catalog No. 50-286-710