Learn More
Aviva Systems Biology Corp SYT12 Antibody - N-terminal region (ARP59489_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59489_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SYT12. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human SYT12. Alternate names: SRG1, SYT11, sytXIIApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-286-718