Learn More
Aviva Systems Biology Corp SYT16 Antibody - N-terminal region (ARP59476_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59476_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SYT16. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human SYT16. Alternate names: CHR14SYT, SYT14L, Strep14, syt14r, yt14rApplicationWB. Species reactivity: Dog, Horse, Human, Rabbit, Rat
Catalog No. 50-286-712