Learn More
Aviva Systems Biology Corp SYT2 Antibody - C-terminal region (ARP59477_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59477_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SYT2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human SYT2. Alternate names: FLJ42519, SytIIApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-713