Learn More
Aviva Systems Biology Corp SYT3 Antibody - N-terminal region (ARP50058_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50058_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SYT3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SYT3. Alternate names: DKFZp761O132, SytIIIApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-60254