Learn More
Aviva Systems Biology Corp SYT5 Antibody - middle region (ARP59484_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59484_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SYT5. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human SYT5. Alternate names: -ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-286-714