Learn More
Aviva Systems Biology Corp SYT7 Antibody - N-terminal region (ARP64596_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64596_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SYT7. It was validated on Western Blot. . Alternate names: IPCA-7, MGC150517, PCANAP7, SYT-VIIApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-288-308