Learn More
Aviva Systems Biology Corp SYTL1 Antibody - N-terminal region (ARP60706_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60706_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SYTL1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human SYTL1. Alternate names: FLJ14996, JFC1, SLP1ApplicationWB. Species reactivity: Human
Catalog No. 50-287-135