Learn More
Aviva Systems Biology Corp SYVN1 Antibody - C-terminal region (ARP43361_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43361_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SYVN1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human SYVN1. Alternate names: HRD1, KIAA1810, MGC40372ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-58508