Learn More
Medchemexpress LLC HY-N1187 1mg Medchemexpress, Syzalterin CAS:94451-48-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN11871MG
Encompass_Preferred
Medchemexpress, HY-N1187 1mg Syzalterin CAS:94451-48-6 Syzalterin is an inhibitor of NO production with an IC50 of 1.87 μg/mL. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2833
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.