Learn More
Ace Glass, Inc. SZ 1 CORK RING 10-100 SZ 1 CORK RING 10-100
SDP
Catalog No. 509558940
Encompass
Size 1 (10-100mL) fine composition cork ring, ideally suited for general laboratory use to support flasks, beakers, dishes, and more. Ring is 30mm thick. OD: 7.6cm, ID: 3.0cm.
Catalog No. 50-955-8940 Supplier Ace Glass, Inc. Supplier No. 1175002
Size 1 (10-100mL) fine composition cork ring, ideally suited for general laboratory use to support flasks, beakers, dishes, and more. Ring is 30mm thick. OD: 7.6cm, ID: 3.0cm.