Learn More
MSC HYFLEX PALM DIP KNIT GLVS
Catalog No. 502738894
Encompass
Series 11-735 Puncture-Resistant Gloves: Size Small, ANSI Cut N/A, Polyurethane, Series 11-735
Catalog No. 50-273-8894 Supplier MSC Supplier No. 36094746
Series 11-735 Puncture-Resistant Gloves: Size Small, ANSI Cut N/A, Polyurethane, Series 11-735