Learn More
Ace Glass, Inc. Cannon-Fenske viscometer, uncalibrated, size 700, 20000-100000 centistokes
SDP
Catalog No. 50975856
Encompass
Cannon-Fenske viscometer, uncalibrated, size 700, 20000-100000 centistokes
Catalog No. 50-975-856 Supplier Ace Glass, Inc. Supplier No. 798829
Cannon-Fenske viscometer, uncalibrated, size 700, 20000-100000 centistokes