Learn More
Ace Glass, Inc. SZ A ADAPTER SZ A ADAPTER
SDP
Catalog No. 50978849
Encompass_Preferred
10mm O.D. glass connecting adapter without flange has a #25 Ace-Thred used for connecting ACE freeze drying vessels to the rubber valves on most other commercially available freeze drying units.
Catalog No. 50-978-849 Supplier Ace Glass, Inc. Supplier No. 669674
10mm O.D. glass connecting adapter without flange has a #25 Ace-Thred used for connecting ACE freeze drying vessels to the rubber valves on most other commercially available freeze drying units.