Learn More
Ace Glass, Inc. 12.7mm O.D. glass connecting adapter with 19.1mm flange
SDP
Catalog No. 50973329
Encompass_Preferred
12.7mm O.D. glass connecting adapter with 19.1mm flange has a #25 Ace-Thred used for connecting ACE freeze drying vessels to the rubber valves on most other commercially available freeze drying units.
Catalog No. 50-973-329 Supplier Ace Glass, Inc. Supplier No. 669676
12.7mm O.D. glass connecting adapter with 19.1mm flange has a #25 Ace-Thred used for connecting ACE freeze drying vessels to the rubber valves on most other commercially available freeze drying units.