Learn More
Ace Glass, Inc. 19.1mm O.D. glass connecting adapter
SDP
Catalog No. 50973330
Encompass_Preferred
19.1mm O.D. glass connecting adapter without flange has a #25 Ace-Thred used for connecting ACE freeze drying vessels to the rubber valves on most other commercially available freeze drying units.
Catalog No. 50-973-330 Supplier Ace Glass, Inc. Supplier No. 669677
19.1mm O.D. glass connecting adapter without flange has a #25 Ace-Thred used for connecting ACE freeze drying vessels to the rubber valves on most other commercially available freeze drying units.