Learn More
MSC EDGE HPPE/NITRILE C/R GLV
Supplier: MSC 80464456
Cut & Puncture Resistant Gloves; Coating Material: Nitrile ; Coating Coverage: Palm & Fingertips ; Glove Type: Abrasion Resistant; Cut & Puncture-Resistant ; ANSI/ISEA Cut Resistance Level: A3