Learn More
Ace Glass, Inc. Joint clip, size 13, Keck; type, all Delrin;, fits 12/5spherical joints, not for positive pressure use, violet, pk/10
SDP
Catalog No. 50975319
Encompass
Joint clip, size 13, Keck; type, all Delrin;, fits 12/5spherical joints, not for positive pressure use, violet, pk/10
Catalog No. 50-975-319 Supplier Ace Glass, Inc. Supplier No. 766812
Joint clip, size 13, Keck; type, all Delrin;, fits 12/5spherical joints, not for positive pressure use, violet, pk/10