Learn More
Ace Glass, Inc. Joint clip, size 19, Keck; type, all Delrin;, fits18/7, 18/9, 7mm & 9mm ID O-ring joints, not for positive pressure use, blue, pk/10
SDP
Catalog No. 50975320
Encompass
Joint clip, size 19, Keck; type, all Delrin;, fits18/7, 18/9, 7mm & 9mm ID O-ring joints, not for positive pressure use, blue, pk/10
Catalog No. 50-975-320 Supplier Ace Glass, Inc. Supplier No. 766818
Joint clip, size 19, Keck; type, all Delrin;, fits18/7, 18/9, 7mm & 9mm ID O-ring joints, not for positive pressure use, blue, pk/10