Learn More
Ace Glass, Inc. SZ2 CORK RING 200-500 SZ2 CORK RING 200-500
SDP
Catalog No. 509558941
Encompass
Size 2 (200-500mL) fine composition cork ring, ideally suited for general laboratory use to support flasks, beakers, dishes, and more. Ring is 30mm thick. OD: 11.1cm, ID: 6.0cm.
Catalog No. 50-955-8941 Supplier Ace Glass, Inc. Supplier No. 1175004
Size 2 (200-500mL) fine composition cork ring, ideally suited for general laboratory use to support flasks, beakers, dishes, and more. Ring is 30mm thick. OD: 11.1cm, ID: 6.0cm.