Learn More
Fine Science Tools Kalt Suture Needles, Stainless Steel, Size: 2
SDP
Supplier:  Fine Science Tools 1205002
Encompass
Product: Kalt Suture Needles - Size 2/ Size: 2/ Needle Diameter: 0.35mm/ Needle Circle: 3/8/ Needle Arch: 16mm/ Needle Chord: 12.5mm/ Needle Alloy: Stainless Steel/ Sterile: No/ Unit: Dozen
Catalog No. NC9876706