Learn More
Ace Glass, Inc. Joint clip, size 29, Keck; type, all Delrin;, fits28/11, 28/15 and 15mm ID O-ring joints, not for positive pressure use, dark red, pk/10
SDP
Catalog No. 50979428
Encompass
Joint clip, size 29, Keck; type, all Delrin;, fits28/11, 28/15 and 15mm ID O-ring joints, not for positive pressure use, dark red, pk/10
Catalog No. 50-979-428 Supplier Ace Glass, Inc. Supplier No. 766828
Joint clip, size 29, Keck; type, all Delrin;, fits28/11, 28/15 and 15mm ID O-ring joints, not for positive pressure use, dark red, pk/10