Learn More
Ace Glass, Inc. SZ3 CORK RING 1000-3000SZ3 CO SZ3 CORK RING 1000-3000SZ3 CO
SDP
Catalog No. 509558942
Encompass
Size 3 (1000-3000mL) fine composition cork ring, ideally suited for general laboratory use to support flasks, beakers, dishes, and more. Ring is 30mm thick. OD: 14.0cm, ID: 8.9cm.
Catalog No. 50-955-8942 Supplier Ace Glass, Inc. Supplier No. 1175006
Size 3 (1000-3000mL) fine composition cork ring, ideally suited for general laboratory use to support flasks, beakers, dishes, and more. Ring is 30mm thick. OD: 14.0cm, ID: 8.9cm.