Learn More
Ace Glass, Inc. SZ4 CORK RING 5000 SZ4 CORK RING 5000
SDP
Catalog No. 509558943
Encompass
Size 4 (5000mL) fine composition cork ring, ideally suited for general laboratory use to support flasks, beakers, dishes, and more. Ring is 30mm thick. OD: 17.1cm, ID: 12.1cm.
Catalog No. 50-955-8943 Supplier Ace Glass, Inc. Supplier No. 1175008
Size 4 (5000mL) fine composition cork ring, ideally suited for general laboratory use to support flasks, beakers, dishes, and more. Ring is 30mm thick. OD: 17.1cm, ID: 12.1cm.