Learn More
Ace Glass, Inc. SZ6 CORK RING 22000-50000 SZ6 CORK RING 22000-50000
SDP
Catalog No. 509558945
Encompass
Size 6 (22,000-50,000mL) fine composition cork ring, ideally suited for general laboratory use to support flasks, beakers, dishes, and more. Ring is 30mm thick. OD: 24.1cm, ID: 18.6cm.
Catalog No. 50-955-8945 Supplier Ace Glass, Inc. Supplier No. 1175012
Size 6 (22,000-50,000mL) fine composition cork ring, ideally suited for general laboratory use to support flasks, beakers, dishes, and more. Ring is 30mm thick. OD: 24.1cm, ID: 18.6cm.