Learn More
MSC EDGE POLYESTR/NITRILE GLV
Catalog No. 502736992
Encompass
Work & General Purpose Gloves; Glove Type: General Purpose ; Lining Material: Polyester ; Cuff Style: Elastic; Knit ; Primary Material: Polyester ; Coating Material: Nitrile ; Coating Coverage: Palm & Fingertips
Catalog No. 50-273-6992 Supplier MSC Supplier No. 80245640
Add to Cart
Add to Cart
Work & General Purpose Gloves; Glove Type: General Purpose ; Lining Material: Polyester ; Cuff Style: Elastic; Knit ; Primary Material: Polyester ; Coating Material: Nitrile ; Coating Coverage: Palm & Fingertips