Learn More
MSC BUTYL CHEM RES GLV
Supplier:  MSC 92992932
Encompass
Chemical Resistant Gloves; Glove Type: General Purpose Chemical Resistant Gloves ; Material: Water Based Butyl ; Size: Small ; Numeric Size: 7 ; Coating Material: Butyl ; Lining Material: Unlined
Catalog No. 50-273-6418