Learn More
MSC PWDR-FREE STERLE DISP GLV
Supplier:  MSC 54245352
Encompass
Series TouchNTuff Disposable Gloves: Size 7, 8.26 mil, Synthetic Polymer-Coated Latex, Cleanroom Grade, Unpowdered
Catalog No. 50-273-8486