Learn More
MSC ALPHATEC CHEM RESIST GLV
Supplier:  MSC 75476978
Encompass
Chemical Resistant Gloves; Glove Type: Type A Chemical Resistant Gloves ; Material: Nylon; Neoprene; Nitrile ; Size: 8 ; Numeric Size: 8 ; Coating Material: Neoprene; Nitrile ; Lining Material: Nylon
Catalog No. 50-273-7086