Learn More
MSC HYFLEX HPPE/NYLON C/R GLV
Catalog No. 502736428
Encompass
Cut & Puncture Resistant Gloves; Coating Material: Nitrile ; Coating Coverage: Palm & Fingertips ; Glove Type: Cut-Resistant; Abrasion Resistant ; ANSI/ISEA Cut Resistance Level: A6 ; Primary Material: Intercept - Tungsten, HPPE, Nylon
Catalog No. 50-273-6428 Supplier MSC Supplier No. 92992064
Add to Cart
Add to Cart
Cut & Puncture Resistant Gloves; Coating Material: Nitrile ; Coating Coverage: Palm & Fingertips ; Glove Type: Cut-Resistant; Abrasion Resistant ; ANSI/ISEA Cut Resistance Level: A6 ; Primary Material: Intercept - Tungsten, HPPE, Nylon