Learn More
Medchemexpress LLC HY-108313 1mg Medchemexpress, T-00127_HEV1 CAS:900874-91-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1083131MG
Encompass_Preferred
Medchemexpress, HY-108313 1mg T-00127_HEV1 CAS:900874-91-1 T-00127_HEV1 is a phosphatidylinositol 4-kinase III beta (PI4KB) inhibitor with an IC50 of 60 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8524