Learn More
TARGETMOL CHEMICALS INC T-0509 5MG
SDP
Catalog No. 502580482
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. T-0509 is a selective beta 1 receptor agonist that increases inotropy in canine ventricular muscle and selectively down-regulates rat cardiac beta 1-adrenoceptors. purity: 98%
Catalog No. 50-258-0482 Supplier TARGETMOL CHEMICALS INC Supplier No. T288955MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. T-0509 is a selective beta 1 receptor agonist that increases inotropy in canine ventricular muscle and selectively down-regulates rat cardiac beta 1-adrenoceptors. purity: 98%