Learn More
J Kem Scientific Inc T-1/16-T12/THERMOCOUPLE
SDP
Catalog No. NC9447905
Encompass
Thermocouples, Blue, T Type, Probe Outer Diameter: 1/16 (in), PTFE Overcoating, Probe Length: 12 (in)
Catalog No. NC9447905 Supplier J Kem Scientific Inc Supplier No. T1/16T12
Thermocouples, Blue, T Type, Probe Outer Diameter: 1/16 (in), PTFE Overcoating, Probe Length: 12 (in)