Learn More
Medchemexpress LLC HY-120356A 5mg Medchemexpress, T-1101 (tosylate) CAS:2250404-95-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY120356A5MG
Encompass_Preferred
Medchemexpress, HY-120356A 5mg T-1101 (tosylate) CAS:2250404-95-4 T-1101 tosylate (TAI-95 tosylate) is a Hec1/Nek2 (Highly expressed in cancer 1 / NIMA-related kinase 2) inhibitor with antitumor activity. T-1101 tosylate is inactive toward normal cells, kinases and hERG Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9876