Learn More
Stellar Scientific Stellar Scientific T-175 Tissue Culture Treated Flask, Vented Cap, Canted Neck, 40/cs
SDP
Catalog No. NC1932996
Encompass
Tissue Culture flasks are manufactured under the most stringent conditions and include a 100% integrity test to assure the reliability with your tissue and cell culture work. Class 100,000 clean room, USP class VI virgin polystyrene, and ISO 9001:2000 quality management systems all contribute to the consistent tissue and cell culture performance of our products. The tissue culture surface treatment used provides a negatively charged hydrophilic surface and results in a consistent cell attachment surface. Growth surfaces are flat and free from striation to maximize usable growth area and transparency.
Catalog No. NC1932996 Supplier Stellar Scientific Supplier No. TC30130
Tissue Culture flasks are manufactured under the most stringent conditions and include a 100% integrity test to assure the reliability with your tissue and cell culture work. Class 100,000 clean room, USP class VI virgin polystyrene, and ISO 9001:2000 quality management systems all contribute to the consistent tissue and cell culture performance of our products. The tissue culture surface treatment used provides a negatively charged hydrophilic surface and results in a consistent cell attachment surface. Growth surfaces are flat and free from striation to maximize usable growth area and transparency.