Learn More
Cayman Chemical T-2 Toxin-13C24, Purity: ≥98%, 1.2 mL
SDP
Catalog No. 502991446
Encompass_Preferred
An internal standard for the quantification of T-2 toxin by GC- or LC-MS
Catalog No. 50-299-1446 Supplier Cayman Chemical Supplier No. 313001.2 ML
An internal standard for the quantification of T-2 toxin by GC- or LC-MS